Safety-Coated Reagent Bottles, Amber, Wide Mouth, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W217958 W217959 W217956 W217957
10847-492CS 121.68 USD
10847-492 10847-494 10847-566 10847-568
Safety-Coated Reagent Bottles, Amber, Wide Mouth, DWK Life Sciences
Bottles Glass Bottles
These amber safety-coated bottles protect light-sensitive samples during storage or field applications.
  • Non-slip coating allows for greater ease and stability when handling wet or dry
  • Autoclaving/sterilizing bottle is not recommended
  • Bottles with caps are used for distilled water, analytical standards, and reagents
  • Metal foil liners are generally more resistant to organic solvents
  • Manufactured from soda lime glass
The strength, slip resistance, and impact absorption qualities of the plastisol safety coating reduce potential breakage and exposure to hazardous materials.
If a bottle is accidentally broken, the PVC based plastisol coating on the outside of bottle retains the contents long enough for proper disposal.
Certificaciones: Conforms to USP Type III requirements.