Puritan® Cary-Blair Transport Medium, Puritan Medical Products
Supplier: Puritan Medical Products
CB-200 CB-500
75999-260CS 100.8 USD
75999-260 75999-262
Puritan® Cary-Blair Transport Medium, Puritan Medical Products
Media for Microbiology Prepared Media for Microbiology
Cary-Blair transport medium for the preservation of fecal specimens containing enteric pathogenic bacteria.
  • 2mL or 5mL Cary-Blair Transport Medium
  • Intended for use in the preservation of specimens containing enteric bacteria
  • Orange cap
Cary-Blair Transport Medium is intended for use in the preservation of clinical fecal and rectal specimens containing enteric bacteria from the collection site to the testing laboratory for examination and culture. For optimum performance, store at 2-25°C.
Certificaciones: Made in USA.
Información para pedidos: Products are final sale.