Tergitol™ TMN-6, Surfactant, Spectrum Chemical
Supplier: Spectrum Chemicals
T1303-100ML T1303-200LTB
700012-616EA 112.8 USD
700012-616 700012-618
Tergitol™ TMN-6, Surfactant, Spectrum Chemical
Cleaning Agents Detergents and Surface Cleansers
Tergitol™ TMN-6, Surfactant is a is a water-soluble, nonionic surfactant that is a superior wetting agent for use in multiple applications including paper & textile processing and hard surface cleaners.
  • 90% solution
  • Superior wetting agent
  • Penetrant & dispersant
  • Narrow gel range
  • Silicone emulsifier
Density at 20°C (68°F): 0.994 g/ml