Silver protein mild

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Silver colloidal, Silver nucleate, Silvol, Mild protargin, Colloidal Silver

SI120-25GM SI120-500GM SI120-5KGBL SI120-100GM SI120-1KGBL
700015-868EA 250.3 USD
700015-868 700015-870 700015-872 700015-864 700015-866
Silver protein mild
Silver protein

Silver Protein, Mild is made of metallic silver particles suspended in a polymer protein solution and is used for its antiseptic properties on cuts and abrasions.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: Ag (with protein)
Storage Temperature: Ambient
MDL Number: MFCD00132268
CAS Number: 9015-51-4

Order Now

Specification Test Results

ASSAY (Ag) 19 - 23 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR