Tergitol™ NP-10, Surfactant, Spectrum
Supplier: Spectrum Chemicals
T1273-20LT T1273-500ML T1273-100ML T1273-4LT
700011-122EA 1869.65 USD
700011-122 700011-126 700011-120 700011-124
Tergitol™ NP-10, Surfactant, Spectrum
Cleaning Agents Detergents and Surface Cleansers
Tergitol™ NP-10 Surfactant is a nonylphenol ethoxylate-based surfactant. It is nonionic and provides excellent detergency, rinse-ability, and low odor. This product is suggested for use in cleaners & degreasers, prewash spotters and metalworking fluids.
  • Excellent detergency
  • Outstanding wetting
  • Versatile solubility characteristics, soluble in water
  • Exceptional handling properties
  • Low odor
  • Excellent rinseability
Density at 20°C (68°F): 1.060 g/ml