Spore Strip, Mesa Labs
Supplier: Mesa Labs
77-01-1-6100 77-01-3-5100 77-01-5-5100T 77-01-3-6100
101384-618PK 223.62 USD
101384-618 101384-620 101384-624 101384-622
Spore Strip, Mesa Labs
Sterilization Indicators
Spore strips are used for sterility testing in various sterilization environments, for both healthcare and industrial applications.
  • Autoclave validations
  • EO Sterilization
  • Used in steam applications
  • Irradiation sterilization
  • Available in multiple species