Ethanol denatured, anhydrous Govt. Form III
Supplier: AQUA SOLUTIONS MS
|
Danger
|
E2438-20L
E2438-4L
E2438-1L
E2438-500ML
101225-504EA
390.22
USD
101225-504
101226-294
101223-528
101223-266
Ethanol denatured, anhydrous Govt. Form III
Ethanol denatured
Formula:
H₃CCH₂OH |
MDL Number:
MFCD00003568 CAS Number: 64-17-5 UN: 1170 ADR: 3,II |
Specification Test Results
Specific Gravity | 0.785 / 0.805 @ 20° C |
Refractive Index | 1.3550 / 1.3750 |
Appearance | Pass |
Gas Chromatograph | Pass |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...