Sodium deoxycholate monohydrate ≥99.0% (by titrimetric analysis), BioXtra

Warning

D5670-25G D5670-5G
220122-136EA 1233.54 USD
220122-136 220122-137
Sodium deoxycholate monohydrate ≥99.0% (by titrimetric analysis), BioXtra
Sodium deoxycholate monohydrate

Sodium deoxycholate (SDC) monohydrate is a bile acid salt used in the solubilization of estrogen synthetase from human placental microsomes.

Formula: C₂₄H₃₉NaO₄·1H₂O
MDL Number: MFCD00150750
CAS Number: 145224-92-6

Order Now

Specification Test Results

pKa 6.58
λmax 310 nm
Extinction Coefficient (EmM) 9.4
Specific Rotation +55 °
pH 7.5 - 9.5 (20 °C, 0.1 M in H₂O)
Solubility H₂O
UV Absorption A260 (0.1 in H₂O) ≤0.10
UV Absorption A280 (0.1 in H₂O) ≤0.08
Sulfate (SO₄²⁻) ≤0.02%
Appearance (Color) White
Appearance (Form) Powder
Solubility (Color) Colorless
Solubility (Turbidity) 0.1 M in H₂O Clear
Insoluble matter passes filter test Pass
Infrared spectrum Conforms to Structure
Sulfate ≤0.02%
Cholic acid (by TLC) ≤0.5%
Chloride Content ≤0.005%
Aluminum (Al) ≤0.002%
Bismuth ( Bi) 0.0005%
Lead (Pb) 0.0005%
Barium 0.0005%
Cadmium (Cd) 0.0005%
Cobalt ( Co) 0.0005%
Strontium (Sr) ≤0.002%
Copper (Cu) 0.0005%
Zinc (Zn) 0.0005%
Calcium (Ca) ≤0.05%
Iron (Fe) ≤0.001%
Lithium ≤0.0005%
Molybdenum (Mo) ≤0.0005%
Nickel ≤0.0005%
Chromium (Cr) ≤0.0005%
Magnesium (Mg) ≤0.002%
Manganese (Mn) ≤0.0005%
Potassium (K) ≤0.010%
Lithium (Li) ≤0.0005%
pH 0.1 M in H₂O at 20 °C 7.5 - 9.5
Specific Rotation, Sodium lamp (c = 2 in H₂O at 20 °C) 42 - 46 °
Purity by Titration ≥99.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR