Zinc undecylenate ≥98.0% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 10-Undecenoic Acid Zinc

ZI125-12KG ZI125-2.5KG ZI125-25KGBL ZI125-500GM
700005-530EA 2903.45 USD
700005-530 95036-668 700005-532 95036-670
Zinc undecylenate ≥98.0% USP
Zinc undecylenate

Zinc Undecylenate, USP is used to treat skin fungal infections such as athlete's foot and jock itch. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: C₂₂H₃₈O₄Zn
Storage Temperature: Ambient
MDL Number: MFCD00067246
CAS Number: 557-08-4

Order Now

Specification Test Results

Assay (Dried Basis) 98.0 - 102.0 %
Loss On Drying max. 1.25 %
Alkalies and Alkaline Earths max. 1.0 %
Identification To Pass Test
Residual Solvents To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR