Glycine zinc salt monohydrate 26%, powder

Supplier: Spectrum Chemical Mfg. Corp.

Z1117-100GM Z1117-500GM Z1117-2.5KG
700016-342EA 93.6 USD
700016-342 700016-346 700016-344
Glycine zinc salt monohydrate 26%, powder
Glycine zinc salt monohydrate

Zinc Glycinate, Monohydrate, 26 Percent, Powder is used as a dietary supplement in some formulations.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂H₅NO₂
Storage Temperature: Ambient
MDL Number: MFCD00070561
CAS Number: 14281-83-5

Order Now

Specification Test Results

Appearance White to Off-Whi
Activity 26% min.
Lead (Pb) max. 10 ppm
Arsenic (As) max. 3 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR