Zinc D-gluconate ≥97.0% USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Zinc gluconate

Z1077-12KG Z1077-2.5KG Z1077-500GM Z1077-50KGBL
700006-262EA 1690.65 USD
700006-262 95036-642 95036-644 75812-632
Zinc D-gluconate ≥97.0% USP
Zinc D-gluconate

ZINC GLUCONATE, USP is a zinc salt of gluconic acid can also be manufactured by fermenting glucose. It comes as a white powder and is used in cold treating lozenges and as one of the more effective ways of delivering zinc as a dietary supplement. Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₁₂H₂₂O₁₄Zn
MW: 455.69 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00868110
CAS Number: 4468-02-4
EINECS: 224-736-9

Order Now

Specification Test Results

Assay (Anhydrous Basis) 97.0 - 102.0 %
pH of a 1 in 100 Solution 5.5 - 7.5
Water max. 11.6 %
Chloride (Cl) max. 0.05 %
Sulfate max. 0.05 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR