Zinc citrate dihydrate ≥97%, Reagent Grade

Supplier: SPECTRUM CHEMICAL MFG CORP

Z1052-100GM Z1052-25GM Z1052-12KG Z1052-500GM Z1052-2.5KG Z1052-50KGBL
700006-222EA 105.07 USD
700006-222 700006-228 700006-224 700006-230 700006-226 700006-232
Zinc citrate dihydrate ≥97%, Reagent Grade
Zinc citrate dihydrate

Zinc Citrate, Dihydrate, Reagent is mainly used in dental care products such as toothpastes, mouthwashes and chewing gums due to its antimicrobial and anti-inflammatory effects and its ability to reduce or inhibit the formation of dental plaque and tartar. In pharmaceuticals, zinc citrate fights common colds. Zinc is increasingly used in food supplements, functional foods and beverages. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formula: C₁₂H₁₄O₁₆Zn₃
MW: 610.368 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150587
CAS Number: 5990-32-9

Order Now

Specification Test Results

Assay 97 % min.
Zinc (Zn) 31 - 33 %
Solubility To Pass Test
Lead (Pb) max. 10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR