Zeolite, Technical Grade

Supplier: SPECTRUM CHEMICAL MFG CORP

ZE110-12KG ZE110-2.5KG ZE110-500GM
700001-900EA 548.41 USD
700001-900 700001-902 700001-904
Zeolite, Technical Grade
Zeolite

Zeolite, Technical is a microporous, aluminosilicate mineral that is often used as an adsorbent and catalyst in the petrochemical industry.


Technical grade products supplied by Spectrum are indicative of a grade suitable for general industrial use.

Formula: Na₂O·Al₂O₃·xSiO₂·yH₂O
Storage Temperature: Ambient
MDL Number: MFCD00132601
CAS Number: 1318-02-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR