Vitamin A palmitate, 1.70 MIU/g USP

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Retinyl palmitate, Retinol palmitate, all-trans-Retinyl palmitate

V1159-100ML V1159-1LT V1159-25ML V1159-5ML V1159-5KGBL
95036-544EA 829.4 USD
95036-544 95036-546 95036-548 95036-550 75812-598
Vitamin A palmitate, 1.70 MIU/g USP
Vitamin A palmitate

Vitamin A Palmitate, 1.70 MIU/g, USP. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₃₆H₆₀O₂
MW: 524.87 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00019414
CAS Number: 79-81-2

Order Now

Specification Test Results

Assay 1.62 MIU/g
Absorbance Ratio @ 325 NM 0.85 min.
Identification To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR