Veegum® R

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Smectite clay minerals

VE110-2.5KG VE110-500GM VE110-12KG VE110-45KGBL
700016-116EA 379.05 USD
700016-116 700016-120 700016-114 700016-118
Veegum® R
Veegum®

Veegum(R) R is Magnesium Aluminum Silicate NF Type IA. It is Vanderbilt Mineral's useful, economical grade for a wide range of applications: pharmaceutical, cosmetic, personal care, veterinary, agricultural, household and industrial products. Typical use levels: Between 0.5% and 3%.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
CAS Number: 12199-37-0

Order Now

Specification Test Results

Viscosity of a 5% Dispersion 225 - 600 cPs

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR