Veegum® HV, granulated

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Smectite clay minerals

V1227-45KGBL V1227-2.5KG V1227-500GM
700016-088EA 1906.25 USD
700016-088 700016-086 700016-090
Veegum® HV, granulated
Veegum®

Veegum(R) HV, Granular is Magnesium Aluminum Silicate NF Type IA. It is Vanderbilt Mineral's useful, economical grade for a wide range of applications: pharmaceutical, cosmetic, personal care, veterinary, agricultural, household and industrial products. Typical use levels: Between 0.5% and 3%.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
CAS Number: 12199-37-0

Order Now

Specification Test Results

Viscosity, BrooKField, 5% Dispersion 8000 - 2200 cP


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR