Vanillyl butyl ether

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 4-(Butoxymethyl)-2-methoxyphenol

V1016-1KG V1016-5KGBL
700003-632EA 1200.7 USD
700003-632 700003-630 700003-634 700003-636
Vanillyl butyl ether
Vanillyl butyl ether

Vanillyl Butyl Ether is an ether of monohydroxybenzoic acid and is added to food products as a flavoring agent. It is also present in cosmetics and personal care products as a fragrance agent.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₂H₁₈O₃
Density: 1.057
Storage Temperature: Ambient
MDL Number: MFCD00238529
CAS Number: 82654-98-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR