Tripropylene glycol (mixture of isomers) ≥97%
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Tripropylene glycol
T1377-3LT
T1377-1LT
T1377-250ML
700003-576EA
235.45
USD
700003-576
700003-572
700003-574
Tripropylene glycol (mixture of isomers) ≥97%
Tripropylene glycol (mixture of isomers)
Tripropylene Glycol is typically used in ink removal formulations and stick deodorant products.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₉H₂₀O₄ MW: 192.26 g/mol Density: 1.021 Storage Temperature: Ambient |
MDL Number:
MFCD00014405 CAS Number: 24800-44-0 |
Specification Test Results
Assay | Approx. 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...