Triethylhexanoin
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Propane-1,2,3-triyl tris(2-ethylhexanoate)
T1338-25GM
T1338-500GM
T1338-100GM
700015-956EA
152.65
USD
700015-956
700015-958
700015-954
Triethylhexanoin
Triethylhexanoin
Triethylhexanoin is used as an antistatic agent and as an emollient in cosmetics and skin conditioning products.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₂₇H₅₀O₆ MW: 470.682 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04038092 CAS Number: 7360-38-5 |
Specification Test Results
Appearance | Slightly Yellow |
Saponification Value | 345.0 - 365.0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...