Triethylene glycol dimethacrylate

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: (Ethane-1,2-diylbis(oxy))bis(ethane-2,1-diyl) bis(2-methylacrylate)

T1453-200LTB T1453-100ML T1453-500ML
700005-426EA 6105.4 USD
700005-426 700005-424 700005-428
Triethylene glycol dimethacrylate
Triethylene glycol dimethacrylate

Triethylene Glycol Dimethacrylate, also known as Triglycol or TEG, is used as a plasticizer for vinyl, as a liquid desiccant for natural gas, and in air conditioning systems; it belongs to a homologous series of dihydroxy alcohols.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₄H₂₂O₆
MW: 286.32 g/mol
Density: 1.092
Storage Temperature: Refrigerator
MDL Number: MFCD00008591
CAS Number: 109-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR