Triethanolamine salicylate ≥85%

Supplier: SPECTRUM CHEMICAL MFG CORP

T1114-25GM T1114-100GM T1114-10KGBL T1114-500GM
700015-878EA 132.3 USD
700015-878 700015-874 700015-876 700015-880
Triethanolamine salicylate ≥85%
Triethanolamine salicylate

Triethanolamine Salicylate is the salt or ester of the colorless organic compound that is known as Triethanolamine and has a viscous texture. It is both a tertiary amine and a triol. It is mainly used as an emulsifier and a surfactant. It neutralizes fatty acids while adjusting and buffering the pH. It is used in the production of cement and has a use in the laboratory as a complexing agent for aluminum ions in aqueous solutions.

Formula: C₆H₁₅NO₃·C₇H₆NO₃
Storage Temperature: Ambient
MDL Number: MFCD00082325
CAS Number: 2174-16-5

Order Now

Specification Test Results

Assay 85 - 95 %
pH of a 5% Aqueous Solution 6.0- 7.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR