1H,1H,2H,2H-Perfluorooctyltriethoxysilane ≥97.5%
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
Triethoxy-1H,1H,2H,2H-tridecafluoro-n-octylsilane, Tridecafluoro-1,1,2,2-tetrahydrooctyl-1-triethoxysilane, Triethoxy(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane;
T1322-25GM
T1322-5GM
700008-262EA
95.63
USD
700008-262
700008-264
700008-266
1H,1H,2H,2H-Perfluorooctyltriethoxysilane ≥97.5%
1H,1H,2H,2H-Perfluorooctyltriethoxysilane
Tridecafluoro-1,1,2,2-tetrahydrooctyl-1-triethoxysilane is used as a binding agent in cosmetic and skin care projects.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₁₄H₁₉F₁₃O₃Si MW: 510.367 g/mol Density: 1.3299 Storage Temperature: Ambient |
MDL Number:
MFCD00042333 CAS Number: 51851-37-7 |
Specification Test Results
Assay | 97.5% min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...