trans-1,4-Dibromo-2-butene ≥98%
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
(E)-1,4-Dibromobut-2-ene, (E)-1,4-Dibromo-2-butene
D1998-25GM
D1998-250GM
10841-986EA
159.85
USD
10841-986
10841-988
trans-1,4-Dibromo-2-butene ≥98%
trans-1,4-Dibromo-2-butene
Storage temperature - REFRIGERATE: Keep tightly closed.
GHS hazard : Corrosion;Skull and crossbones
UN2923
Precaución: Storage temperature - REFRIGERATE: Keep tightly closed.
Formula:
C₄H₆Br₂ MW: 213.9 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00000249 CAS Number: 821-06-7 UN: 2923 ADR: 86.1,II |
Specification Test Results
Purity (GC) | 98 % |
Melting Point | 50.0 - 54.0°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...