Thyme oil, red

Supplier: Spectrum Chemical Mfg. Corp.

T1047-100KGB T1047-100ML T1047-500MLG
700010-828EA 67525.3 USD
700010-828 700010-830 700010-832
Thyme oil, red
Thyme oil

Thyme Oil, Red, an essential oil of thyme, has many applications including as an antiseptic and as an effective agent against certain fungi.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Density: 0.917
Storage Temperature: Ambient
MDL Number: MFCD00148640
CAS Number: 8007-46-3
UN: 2924
ADR: 38,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR