Thiamine nitrate ≥98.0% FCC
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Vitamin B1 Nitrate
T1054-25GM
T1054-1KG
T1054-500GM
T1054-100GM
T1054-25KGBL
95036-282EA
83.5
USD
95036-282
95036-280
95036-284
95036-278
75812-524
Thiamine nitrate ≥98.0% FCC
Thiamine nitrate
Thiamine Mononitrate, FCC is in the B vitamin group and is water soluble. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
C₁₂H₁₇N₅O₄S MW: 327.36 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00036330 CAS Number: 532-43-4 |
Specification Test Results
Assay (Dried Basis) | 98.0 - 102.0 % |
pH of a 1 in 50 Solution | 6.0 - 7.5 |
Chloride (Cl) | max. 0.06 % |
Lead (Pb) | max. 2 mg/kg |
Loss On Drying | max. 1.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...