Tetrabutylammonium dihydrogen phosphate ≥99.0% for HPLC

Supplier: Spectrum Chemical Mfg. Corp.

T2329-5GM T2329-1GM T2329-1KGBL T2329-25GM
700007-672EA 364.59 USD
700007-672 700007-666 700007-668 700007-670
Tetrabutylammonium dihydrogen phosphate ≥99.0% for HPLC
Tetrabutylammonium dihydrogen phosphate

Tetrabutylammonium Dihydrogen Phosphate, HPLC Grade is a chemical compound and often used as a solution in water or alcohol and serves as a buffering reagent in HPLC.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₆H₃₈NO₄P
Storage Temperature: Ambient
MDL Number: MFCD00064526
CAS Number: 5574-97-0

Order Now

Specification Test Results

Assay 99.0 Wt % min.
Absorbance To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR