Terpin monohydrate ≥98.0% USP

Supplier: Spectrum Chemical Mfg. Corp.

TE110-100GM TE110-500GM TE110-45KGBL TE110-2.5KG
95036-228EA 165.5 USD
95036-228 95036-232 700003-134 95036-230
Terpin monohydrate ≥98.0% USP
Terpin monohydrate

TERPIN, HYDRATE, USP, an expectorant, is used in the treatment of acute and chronic bronchitis. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₁₀H₂₀O₂
Storage Temperature: Ambient
MDL Number: MFCD00149225
CAS Number: 2451-01-6

Order Now

Specification Test Results

Assay (C₁₀H₂₀O₂) 98.0 - 100.5 %
Water 9.0 - 10.0 %
Residue On Ignition max. 0.1 %
Residual Turpentine To Pass Test
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR