Tergitol® TMN-10 surfactant

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Isolaureth-10, Polyethylene glycol trimethylnonyl ether

T1288-100ML T1288-20LT T1288-4LT T1288-500ML
700012-608EA 120.1 USD
700012-608 700012-610 700012-612 700012-614
Tergitol® TMN-10 surfactant
Tergitol® TMN-10

Tergitol(R) TMN-10, Surfactant is a Nonylphenol Ethoxylate. It is nonionic and provides excellent detergency, outstanding wetting, versatile solubility characteristics and low odor. This product is suggested for use in cleaners & degreasers, prewash spotters and oil field chemicals


Used in cleaners & degreasers, prewash spotters and oil field chemicals.

Formula: C₁₂H₂₅O(CH₂CH₂O)nH
MW: ∼ 540 g/mol
Density: 1.042
Storage Temperature: Ambient
MDL Number: MFCD00132411
CAS Number: 60828-78-6

Order Now

Specification Test Results

Cloud Point of a 1 % Aqueous Solution 72°- 80°C
pH of a 10% Solution @ 25°C 5.0 - 8.0
Water 8 - 10 %
Suspended Matter Substantially Fr
Color (APHA) max. 200

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR