Sulfaquinoxaline sodium salt ≥95%

Supplier: Spectrum Chemical Mfg. Corp.

S2054-100GM S2054-25GM S2054-25KGBL S2054-500GM S2054-5GM
700009-268EA 379.9 USD
700009-268 700009-270 700009-272 700009-274 700009-276
Sulfaquinoxaline sodium salt ≥95%
Sulfaquinoxaline sodium salt

Sulfaquinoxaline Sodium Salt, also known as 4-Amino-N-2-quinoxalinylbenzenesulfonamide, is a veterinary medicine used to treat coccidiosis in sheep and cattle.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₄H₁₁N₄NaO₂S
Storage Temperature: Ambient
MDL Number: MFCD00799608
CAS Number: 967-80-6

Order Now

Specification Test Results

Assay Approx. 95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR