Stevioside
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
(2S,3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl (4R,4aS,6aR,9S,11aR,11bS)-9-(((2S,3R,4S,5S,6R)-4,5-dihydroxy-6-(hydroxymethyl)-3-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-4,11b-dimethyl-8-methylenetetradecahydro-6a,9-methanocyclohepta[a]naphthalene-4-carboxylate
Stevioside, a glycoside, comes from the stevia plant and is used as a sweetener.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
C₃₈H₆₀O₁₈ MW: 804.88 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00079561 CAS Number: 57817-89-7 |
Specification Test Results
Steviol Glycosides (Dried Basis) | 80 % |
Loss On Drying | max.5.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...