Sorbic acid ≥99.0%, powder FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: (E,E)-2,4-Hexadienoic acid, (2E,4E)-2,4-Hexadienoic acid, trans,trans-2,4-Hexadienoic acid, 2,4-Hexadienoic acid, (2E,4E)-Hexa-2,4-dienoic acid

S1517-2.5KG S1517-500GM S1517-12KG
95035-998EA 607.75 USD
95035-998 95036-000 75812-342
Sorbic acid ≥99.0%, powder FCC
Sorbic acid

Sorbic Acid, Powder, FCC is used as a food preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark

Formula: CH₃CH=CHCH=CHCOOH
MW: 112.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002703
CAS Number: 110-44-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=440c50b4fc52d9ff0b52afa7ba596c860f95e0e7&vid=f9b0ff409010208781696e9ca3314c02aaf1af36&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Anhydrous Basis) 99.0 - 101.0 %
Melting Range 132 - 135 °C
Lead (Pb) max. 2 mg/kg
Residue On Ignition max. 0.2 %
Water max. 0.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR