Sodium ricinolate

Supplier: Spectrum Chemical Mfg. Corp.

S1164-100GM S1164-25GM
700015-616EA 466.7 USD
700015-616 700015-618
Sodium ricinolate
Sodium ricinolate

Sodium Ricinoleate, also known as Ricinoleic acid sodium salt, is the sodium salt of ricinoleic acid and fatty acid from castor oil; it has applications in soap-making and as a bactericide.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₈H₃₄O₃
Storage Temperature: Ambient
MDL Number: MFCD00050760
CAS Number: 5323-95-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR