Sodium perborate tetrahydrate, granulated, purified

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Sodium peroxometaborate tetrahydrate

S1375-12KG S1375-500GM S1375-50KGBL S1375-125GM S1375-2.5KG
700001-908EA 894.1 USD
700001-908 700001-912 700001-914 700001-906 700001-910
Sodium perborate tetrahydrate, granulated, purified
Sodium perborate tetrahydrate

Sodium Perborate, Tetrahydrate, Granular, Purified is used as a bleaching agent or an antiseptic. It is also known to crystallize as the monohydrate. The Purified Grade describes chemicals of good quality where there are no official standards and is normally limited to inorganic chemicals.


GHS hazard : Flame over circle;Exclamation mark;Health hazard
UN1479

Formula: NaH₂BO₄·4H₂O
MW: 171.87 g/mol
Melting Pt: 60
Storage Temperature: Ambient
MDL Number: MFCD00149231
CAS Number: 13517-20-9
UN: 1479
ADR: 5.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR