2-Phenylphenol sodium salt tetrahydrate, Technical Grade

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 2-Hydroxybiphenyl Sodium Salt, Sodium 2-Phenylphenolate, Sodium 2-Hydroxybiphenyl, 2-Biphenylol Sodium Salt, Sodium o-Phenylphenate, Sodium 2-Biphenylolate

S1156-1KG S1156-250GM
700003-256EA 431.45 USD
700003-256 700003-258
2-Phenylphenol sodium salt tetrahydrate, Technical Grade
2-Phenylphenol sodium salt tetrahydrate

Sodium o-Phenylphenate, Tetrahydrate, Technical, also known as biphenylol or 2-Phenylphenol, is an organic compound and a biocide used as an agricultural fungicide.


Technical grade products supplied by Spectrum are indicative of a grade suitable for general industrial use.


Precaución: Hygroscopic: Keep tightly closed.

Formula: C₁₂H₁₀O
MW: 192.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00002209
CAS Number: 6152-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR