Sodium ligninsulfonate, Technical Grade
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Ligninsulphonic acid Sodium Salt
S1854-100GM
S1854-2.5KG
S1854-500GM
700011-532EA
143.94
USD
700011-532
700011-534
700011-536
Sodium ligninsulfonate, Technical Grade
Sodium ligninsulfonate
Sodium Lignosulfonate, Technical, or lignosulfonic acid, has antimicrobial and preservative properties.
Technical grade products supplied by Spectrum are indicative of a grade suitable for general industrial use.
Storage Temperature:
Ambient |
MDL Number:
MFCD00135572 CAS Number: 8061-51-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...