Sodium isoascorbate monohydrate ≥98.0% FCC

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Isoascorbic acid Sodium salt, Sodium Erythorbate, Sodium (R)-2-((R)-1,2-dihydroxyethyl)-4-hydroxy-5-oxo-2,5-dihydrofuran-3-olate, Erythorbic acid Sodium salt, Sodium D-Araboascorbate, D-Araboascorbic acid Sodium salt

S1223-12KG S1223-125GM S1223-2.5KG S1223-500GM
700015-696EA 2381.1 USD
700015-696 95035-810 95035-812 95035-814
Sodium isoascorbate monohydrate ≥98.0% FCC
Sodium isoascorbate monohydrate

Sodium Erythorbate, FCC is used as an antioxidant preservative. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₆H₈O₆
MW: 198.11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00067298
CAS Number: 6381-77-7

Order Now

Specification Test Results

Assay 98.0 - 100.5 %
Optical Rotation +95.5 to +98.0
Lead (Pb) max. 5 mg/kg
Loss On Drying max. 0.25 %
Oxalate (C₂O₄) To Pass Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR