Sodium hydrogen carbonate ≥99.0%, powder FCC
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Sodium bicarbonate
S1148-2.5CS4
S1148-500GM
S1148-2.5KG
S1148-500CS6
S1148-12KG
S1148-45KGBL
95035-714CS
784.9
USD
95035-714
95035-720
95035-716
95035-718
75812-170
75812-172
Sodium hydrogen carbonate ≥99.0%, powder FCC
Sodium hydrogen carbonate
Sodium Bicarbonate, Powder, FCC is used as a leavening agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
Formula:
NaHCO₃ MW: 84.01 g/mol Melting Pt: 270 Storage Temperature: Ambient |
MDL Number:
MFCD00003528 CAS Number: 144-55-8 Merck Index: 13,08655 |
|
Specification Test Results
Assay (Dried Basis) | 99.0 - 100.5 % |
Ammonia (NH₃) | To Pass Test |
Insoluble Substances | To Pass Test |
Lead (Pb) | max. 2 mg/kg |
Loss On Drying | max. 0.25 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...