Sodium L(+)-ascorbate ≥99.0% USP
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Sodium ascorbate, L(+)-Ascorbic acid sodium salt, L-Ascorbic acid sodium salt, Sodium (R)-2-((S)-1,2-dihydroxyethyl)-4-hydroxy-5-oxo-2,5-dihydrofuran-3-olate, Acidum ascorbicum sodium salt
SO108-100GM
SO108-500GM
SO108-5KG
SO108-50KGBL
95035-688EA
115.56
USD
95035-688
95035-690
700009-378
75812-420
Sodium L(+)-ascorbate ≥99.0% USP
Sodium L(+)-ascorbate
Sodium Ascorbate, USP is one of a number of mineral salts of ascorbic acid (vitamin C).
All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formula:
C₆H₇NaO₆ MW: 198.11 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00082340 CAS Number: 134-03-2 |
Specification Test Results
Assay (C₆H₇Nao₆) | 99.0 - 101.0 % |
pH of a 10% Solution | 7.0 - 8.0 |
Specific Rotation [A]D25 | +103° to +108° |
Loss On Drying | max. 0.25 % |
Heavy Metals | max. 0.002 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...