S-(Carboxymethyl)-L-cysteine

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 3-[(Carboxymethyl)thio]alanine, Carbocisteine, Carbocysteine

C3071-100GM C3071-25KGBL
700007-252EA 322.95 USD
700007-252 700007-256
S-(Carboxymethyl)-L-cysteine
S-(Carboxymethyl)-L-cysteine

S-Carboxymethyl-L-cysteine, also known as Rhinathiol or Humex, is a mucolytic that minimizes the viscosity of sputum so it can be used to treat chronic obstructive pulmonary disorder and bronchiectasis.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₅H₉NO₄S
MW: 179.19 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00002614
CAS Number: 638-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR