Saccharin ≥99.0%, powder NF
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
o-Sulphobenzimide, o-Benzoic sulphimide
SA105-12KGBL
SA105-125GM
SA105-2.5KG
SA105-500GM
89078-988EA
4650.7
USD
89078-988
95035-592
95035-594
95035-596
Saccharin ≥99.0%, powder NF
Saccharin
Saccharin, Powder, NF is a pharmaceutical grade substance whose most common use is as an artificial sweetener. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
Formula:
C₇H₅NO₃S MW: 183.19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00005866 CAS Number: 81-07-2 |
Specification Test Results
Assay (C₇H₅NO₃S) | 99.0 - 101.0 % |
Clarity of Solution | To Pass Test |
Color of Solution | To Pass Test |
Melting Range | 226 - 230 °C |
Loss On Drying | max. 1.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...