Rutin, powder

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Ilixanthi, Myrticalorin, Globulariacitrin, Osyritrin, 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4-oxo-4H-chromen-3-yl, Quercetin 3-rutinoside, Rutinoside, 3-Rutinosylquercetin, Quercetin 3-rhamnoglucoside, Paliuroside, 2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-3-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-((((2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)methyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one, 3,3′,4′,5,7-Pentahydroxyflavone 3-rutinoside, Tanrutin, Phytomelin, Violaquercitrin, Sophorin, Rutoside

RU105-25GM RU105-25KGBL RU105-100GM RU105-500GM
700015-568EA 215.38 USD
700015-568 700015-570 700015-566 700015-572
Rutin, powder
Rutin

Rutin, Powder is also known as sophorin or rutoside and is the glycoside that is between the disaccharide rutinose and flavonol quercetin. It is one of the phenolic compounds found in certain plant species that give it its antioxidant properties.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₇H₃₀O₁₆
MW: 610.521 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00006830
CAS Number: 153-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR