Rose Bengal

Supplier: SPECTRUM CHEMICAL MFG CORP

RO115-25GM RO115-500GMB RO115-10GM
700010-148EA 200.68 USD
700010-148 700010-150 700010-146
Rose Bengal
Rose Bengal

Rose Bengal can be used in synthetic chemistry to generate singlet oxygen from triplet oxygen.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₂₀H₂Cl₄I₄O₅Na₂
Storage Temperature: Ambient
MDL Number: MFCD00151169
CAS Number: 632-69-9

Order Now

Specification Test Results

Absorption Max Approx. 548 NM


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR