Resorcinol monoacetate
Supplier: Spectrum Chemical Mfg. Corp.
|
|
Resorcinol Monoacetate is an organic compound formed by the combination of resorcinol and acetic acid. In cosmetics and personal care product formulations, it functions as an anti-dandruff agent, anti-aging agent and conditioning agent in body, hair and skin products. It is used in topical products as an anti-acne ingredient in OTC drug products. It appears as an amber, oily liquid. It is partially soluble in cold water
GHS hazard : Exclamation mark
Formula:
C₈H₈O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00002266 CAS Number: 102-29-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...