Quercetin dihydrate ≥95%

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one dihydrate

Q2150-100GM Q2150-2.5KG Q2150-25GM Q2150-500GM
10841-964EA 306.1 USD
10841-964 10841-966 10841-968 10841-970
Quercetin dihydrate ≥95%
Quercetin dihydrate

Quercetin, Dihydrate, also known as meletin, is a flavonol found in several types of vegetables, fruits, leaves and grains.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₅H₁₄O₉
MW: 338.268 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149487
CAS Number: 6151-25-3
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay (C₁₅H₁₀O₇.2H₂O) 95 % min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR