Potassium sorbate ≥98.0%, granulated NF

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Potassium 2,4-Hexadienoate, Sorbic acid potassium salt, Potassium (2E,4E)-hexa-2,4-dienoate

PO301-12KG PO301-2.5KG PO301-125GM PO301-500GM
700005-520EA 1722.55 USD
700005-520 95035-218 95035-216 95035-220
Potassium sorbate ≥98.0%, granulated NF
Potassium sorbate

Potassium Sorbate, Granular, NF is used as an antimicrobial preservative. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


GHS hazard : Exclamation mark

Formula: CH₃CH=CHCH=CHCOOK
MW: 150.22 g/mol
Melting Pt: 270
Storage Temperature: Ambient
MDL Number: MFCD00016546
CAS Number: 24634-61-5
Merck Index: 13,07756
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=500421971592bd8f37c1a6b88f97c1de033cbf87&vid=e8c4a1d0da78946f174af6a94e5c78fc9c16de05&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Dried Basis) 98.0 - 101.0 %
Acidity or Alkalinity To Pass Test
Loss On Drying max. 1.0 %
Heavy Metals max. 10 ppm
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR