Potassium disulfite ≥90.0%, crystals FCC

Supplier: Spectrum Chemical Mfg. Corp.
Warning

PO191-12KG PO191-2.5KG PO191-50KGBL
75812-058EA 1139.65 USD
75812-058 75812-060 75812-062
Potassium disulfite ≥90.0%, crystals FCC
Potassium disulphite

Potassium Metabisulfite, Crystal, FCC is used as a food preservative and an antioxidant. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


Storage temperature - AIR SENSITIVE, HEAT SENSITIVE: Keep tightly closed in a cool place.

Formula: K₂O₅S₂
Storage Temperature: Ambient
MDL Number: MFCD00167605
CAS Number: 16731-55-8

Order Now

Specification Test Results

Assay (K₂S₂O₅) 90.0 % min.
Lead (Pb) max.2 mg/kg
Iron (Fe) max. 10 mg/kg
Selenium (Se) max. 5 mg/kg
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR