Ponceau 3R

Supplier: SPECTRUM CHEMICAL MFG CORP

PO144-5GM PO144-25GM
700015-496EA 130.2 USD
700015-496 700015-494
Ponceau 3R
Ponceau 3R

Ponceau 3R is a dye for wool, colorant in externally applied drugs and cosmetics, colorant in foods and biological stain.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
MDL Number: MFCD00036432
CAS Number: 3564-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR