Ponceau Xylidine

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Acid Red 26, Xylidine Ponceau 2R, Xylidine ponceau, Ponceau 2R, CI 16150

PO143-25GM PO143-5GM
700015-490EA 293.05 USD
700015-490 700015-492
Ponceau Xylidine
Ponceau Xylidine

Ponceau 2R, also known as xylidine ponceau or Red R, is used in histology for staining.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₈H₁₆N₂O₇S₂
Storage Temperature: Ambient
MDL Number: MFCD00003897
CAS Number: 3761-53-3

Order Now

Specification Test Results

Absorption Max (In Water) Approx. 503 (388


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR