Polyethylene glycol monostearate 40 NF Type II

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Polyoxyethylene stearate, Polyoxyl stearate

P1159-500GM P1159-12KG P1159-45KGBL P1159-2.5KG P1159-113KGBL
95035-022EA 202.75 USD
95035-022 700010-992 700010-994 95035-020 75811-798
Polyethylene glycol monostearate 40 NF Type II
Polyethylene glycol monostearate

Polyoxyl 40 Stearate, Type II, NF is commonly used in pharmaceutical formulations as an emulsifier and a surfactant. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: HO(CH₂CH₂O)nOCC₁₇H₃₅
Storage Temperature: Ambient
MDL Number: MFCD00148007
CAS Number: 9004-99-3

Order Now

Specification Test Results

Stearic Acid and Palmitic Acid 96.0 % min.-
Stearic Acid 90.0 - 99.0 %
Free Polyethylene Glycols 17 - 27 %
Alkalinity To Pass Test
Hydroxyl Value 25 - 40


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR