Castor oil, hydrogenated, ethoxylated 60, hydrogenated
Supplier: Spectrum Chemical Mfg. Corp.
Polyethyleneglycol 60 Castor Oil, Hydrogenated is a polyether compound that is used in a wide variety of fields including pharmaceutical manufacturing as an excipient and active ingredient. Due to its low toxicity it can be used as a lubricating coating for various surfaces in aqueous and non-aqueous environments, a reagent in biochemistry to create very high osmotic pressures, a polar stationary phase for gas chromatography and as a binder.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Storage Temperature:
Ambient |
MDL Number:
MFCD01771439 CAS Number: 61788-85-0 |
Specification Test Results
Appearance @ 25° C | Off-White Solid |
Saponification Number | 40 - 50 |
Hydroxyl Value | 50 - 70 |
Moisture (KF) | max. 2% |
Acid Value | max. 1.5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...